Hydrochloric acid 32%, TECHNICAL
Supplier: VWR Chemicals
20256.290EA
0
EUR
20256.290
Hydrochloric acid 32%, TECHNICAL
Hydrochloric acid
Formula:
HCl MW: 36,46 g/mol Boiling Pt: 110 °C (1013 hPa) Melting Pt: –30 °C Density: 1,18 g/cm³ (20 °C) |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 UN: 1789 ADR: 8,II REACH: 01-2119484862-27 |
Specification Test Results
Assay | 31 - 33 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...