Hydrochloric acid standardised solution 5.0 N
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Hydrochloric acid solution, Muriatic acid
35638.K2 35638.K7
35638.K2EA 44.5 EUR
35638.K2 35638.K7
Hydrochloric acid standardised solution 5.0 N
Hydrochloric acid
CAS No.: 7647-01-0 Molecular Formula: HCl Formula Weight: 36.46 Physical Form: Liquid Appearance: Colourless to pale yellow UN No.: 1789 MDL No.: MFCD00011324
Formula: HCl
MW: 36.46 g/mol
Density: 1.06 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II