VWR® Volumetric Flasks, Class A, Glass, Clear, NS Plastic Stopper
Supplier: VWR
VWR®
612-3741EA 23.1 EUR
612-3741 612-3740 612-3819 612-3818 612-3746 612-3738 612-3743 612-3820 612-3742 612-3745 612-3744
VWR® Volumetric Flasks, Class A, Glass, Clear, NS Plastic Stopper
Flasks Volumetric Flasks
Borosilicate 3.3 glass, class A.
  • PP white stopper
  • Marks and inscriptions in blue enamel
  • Calibrated to contain (TC, In)
Certifikace: DIN EN ISO 1042
Informace pro objednávání: Batch number and batch certificate included.