Orthophosphoric acid 85%
Supplier: Roth Carl
Danger
Synonyms: Phosphoric acid
9079.1 9079.2
ROTH9079.1EA 0 EUR
ROTH9079.1 ROTH9079.2
Orthophosphoric acid 85%
Orthophosphoric acid
Formula: H₃PO₄
MW: 98 g/mol
Boiling Pt: 158
Melting Pt: 21
Density: 1.71…1.87 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8 III
Merck Index: 14,07344