(±)-Epichlorhydrin ≥99%
Supplier: Thermo Scientific
Synonyms:
(±)-1-Chloro-2,3-epoxypropane, Epichlorohydrin, (±)-3-Chloro-1,2-epoxypropane, 1-Chloro-2,3-epoxypropane, Epichlorhydrin, (±)-Glycerol epichlorhydrin, 3-Chloro-1,2-epoxypropane
Purity: 99%
CAS No.: 106-89-8
Molecular Formula: C3H5ClO
Formula Weight: 92.53
Flash Point: 32°(90°F)
Physical Form: Liquid
Density: 1.182
Refractive Index: 1.4380
Appearance: Colourless
Odour: Like chlorine
Melting Point: -57°
Boiling Point: 115-117°
Beilstein Reference: 79785
Merck Reference: 14,3611
UN No.: 2023
MDL No.: MFCD00005132
A cyclodextrin cross-linking reagent
Formula:
C₃H₅ClO MW: 92.52 g/mol |
MDL Number:
MFCD00005132 CAS Number: 106-89-8 Merck Index: 14,3611 |