DL-Camphor
Supplier: Thermo Scientific
MAYBJFD03998.250EA
0
EUR
MAYBJFD03998.250
MAYBJFD03998.1
DL-Camphor
DL-Camphor
Formula:
C₁₀H₁₆O MW: 152,24 g/mol Boiling Pt: 204 °C (1013 hPa) Melting Pt: ∼180 °C Density: 0,992 g/cm³ (25 °C) Flash Pt: 66 °C |
MDL Number:
MFCD00074738 UN: 2717 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...