DL-Camphor racemate Ph. Eur., BP, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: (±)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, Camphor, 1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one, DL-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, (±)-1,7,7-Trimethyl-2-norbornanone

M-Clarity™ quality level = MQ200

20506-25KG 20506-500G 20506-2KG
SIAL20506-25KGEA 1560 EUR
SIAL20506-25KG SIAL20506-500G SIAL20506-2KG
DL-Camphor racemate Ph. Eur., BP, Sigma-Aldrich®
DL-Camphor
Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Boiling Pt: 204 °C (1013 hPa)
Melting Pt: ∼180 °C
Density: 0.992 g/cm³ (25 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00074738
CAS Number: 76-22-2
EINECS: 200-945-0
UN: 2717
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR