DL-Camphor, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
21310-1KG-F 21310-250G-F 21310-50G-F
SIAL21310-1KG-FEA 96.5 EUR
SIAL21310-1KG-F SIAL21310-250G-F SIAL21310-50G-F
DL-Camphor, Sigma-Aldrich®
DL-Camphor
Formula: C₁₀H₁₆O
Boiling Pt: 204 °C (1013 hPa)
Melting Pt: ∼180 °C
Density: 0.992 g/cm³ (25 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00074738
CAS Number: 76-22-2
EINECS: 200-945-0
UN: 2717
ADR: 4.1,III