Potassium chloride 3 mol/l (3 N) in aqueous solution
Supplier: SI Analytics
Synonyms: Potassiumchloride
285138554 285138427
SCOI285138554EA 58 EUR
SCOI285138554 662-6185
Potassium chloride 3 mol/l (3 N) in aqueous solution
Potassium chloride
Formula: KCl
MW: 74.55 g/mol
Boiling Pt: 1420
Melting Pt: 773
Density: 1.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621