Hydrochloric acid 0.1 mol/l (0.1 N), BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Synonyms: Hydrochloric acid solution, Muriatic acid
7038.1000EA 35.71 EUR
7038.1000 7038.9020
Hydrochloric acid 0.1 mol/l (0.1 N), BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Hydrochloric acid
Formula: HCl
MW: 36.46 g/mol
Density: 1.004 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 III
Order Now
Specification Test Results
Titer (mol/l) 0.0997 - 0.1003
Trace Impurities (in ppm)
Ammonium (NH4) max. 3
Iron (Fe) max. 0.1
Heavy Metals (as Pb) max. 0.5
Sulfate (SO4) max. 1