Hydrochloric acid 0.5 mol/l (0.5 N), AVS TITRINORM Reag. Ph. Eur. Volumetric
Supplier: VWR Chemicals
Warning
Synonyms: Hydrochloric acid solution, Muriatic acid
31954.290EA 19.8 EUR
31954.290 31954.368 31954.404
Hydrochloric acid 0.5 mol/l (0.5 N), AVS TITRINORM Reag. Ph. Eur. Volumetric
Hydrochloric acid
Formula: HCl
MW: 36.46 g/mol
Density: 1.02 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 III
Order Now
Specification Test Results
Titer (20°C; real value 0.2 % accuracy) 0.499 - 0.501 mol/l
NIST traceable Confirmed