Counting chambers, BLAUBRAND® Bürker-Türk
Supplier: Brand
719505 719520
BRND719505EA 59 EUR
BRND719505 631-1159
Counting chambers, BLAUBRAND® Bürker-Türk
Slides Counting Chambers
Large squares are subdivided into 16 group squares of 0,2 mm sides.
  • Ruling shows nine large squares of 1 mm²
  • Central large square with sub square groups
  • 16 mini squares with 0,05 mm sides (= 0,0025 mm²)