Cover glasses for counting chambers
Supplier: Brand
723015 723014 723016
631-1190EA 17.5 EUR
631-1190 631-1193 631-1192
Cover glasses for counting chambers
Cover Glasses
These hæmocytometer cover glasses differ from ordinary cover glasses by their plain ground and polished surface.
  • Refractive index ne = 1,52 ±0,01
  • Abbe number ve = 56,5 ±0,5
  • Flatness tolerance ±3 µm