Cover glasses
Supplier: Brand
470045 470050 470055 470060 470816 470819 470820
BRND470045EA 61.6 EUR
BRND470045 BRND470050 BRND470055 BRND470060 BRND470816 BRND470819 BRND470820
Cover glasses
Cover Glasses
These cover glasses are made of pure white clear glass and offer good chemical resistance.
  • Automatic production guarantees ready to use quality
  • Clean, dust and grease free
  • Distortion-free
Flatness: ±3 µm