(S)-(+)-2-Chloro-1-propanol, Sigma-Aldrich®
Supplier: Merck
Synonyms: (S)-2-Chloropropan-1-ol
M-Clarity™ quality level = MQ100
296708-1G 296708-250MG
SIAL296708-1GEA 394 EUR
SIAL296708-1G SIAL296708-250MG
(S)-(+)-2-Chloro-1-propanol, Sigma-Aldrich®
(S)-(+)-2-Chloro-1-propanol
Formula: H₃CCHClCH₂OH
MW: 94.54 g/mol
Boiling Pt: 124
Density: 1.1025 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010812
CAS Number: 19210-21-0
UN: 2611
ADR: 6.1 II