Lithium hydroxide monohydrate, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
254274-10G 254274-250G 254274-50G
SIAL254274-10GEA 56.5 EUR
SIAL254274-10G SIAL254274-250G SIAL254274-50G
Lithium hydroxide monohydrate, Sigma-Aldrich®
Lithium hydroxide monohydrate
Formula: LiOH·1H₂O
MW: 41.96 g/mol
Boiling Pt: 920
Melting Pt: 462
Density: 1.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149772
CAS Number: 1310-66-3
EINECS: 215-183-4
UN: 2680
ADR: 8 II
Merck Index: 13,05556