Disodium hydrogen phosphate dodecahydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: sec-Sodium phosphate, Sodium hydrogen phosphate dodecahydrate, Disodium phosphate, Dibasic sodium phosphate dodecahydrate, Sodium phosphate dibasic dodecahydrate, di-Sodium hydrogen phosphate dodecahydrate, Sodium monohydrogen phosphate dodecahydrate, di-Sodium hydrogen orthophosphate dodecahydrate
71649-1KG 71649-500G
SIAL71649-1KGEA 152 EUR
SIAL71649-1KG SIAL71649-500G
Disodium hydrogen phosphate dodecahydrate, Sigma-Aldrich®
di-Sodium hydrogen phosphate dodecahydrate
Formula: Na₂HPO₄·12H₂O
MW: 358.14 g/mol
Melting Pt: 35
Density: 1.52 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149181
CAS Number: 10039-32-4
EINECS: 231-448-7
Merck Index: 13,08733