Disodium hydrogen phosphate dihydrate, Sigma-Aldrich®
Supplier: Merck
71643-1KG 71643-250G
SIAL71643-1KGEA 169 EUR
SIAL71643-1KG SIAL71643-250G
Disodium hydrogen phosphate dihydrate, Sigma-Aldrich®
di-Sodium hydrogen phosphate dihydrate
Formula: Na₂HPO₄·2H₂O
Melting Pt: 92.5 °C
Density: 2.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149182
CAS Number: 10028-24-7
EINECS: 231-448-7