Sodium dihydrogen phosphate dihydrate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Phosphoric acid (sodium hydrate), Sodium phosphate monobasic dihydrate, Sodium dihydrogen orthophosphate dihydrate

71505-1KG 71505-250G
SIAL71505-1KGEA 182 EUR
SIAL71505-1KG SIAL71505-250G
Sodium dihydrogen phosphate dihydrate, Sigma-Aldrich®
Sodium dihydrogen phosphate dihydrate
Formula: NaH₂PO₄·2H₂O
MW: 156.01 g/mol
Boiling Pt: ∼650 °C (1013 hPa)
Melting Pt: 60 °C
Density: 1.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149209
CAS Number: 13472-35-0
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 12,08806

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR