Sodium thiosulphate pentahydrate
Supplier: Roth Carl
P034.1 P034.2 P034.5
ROTHP034.1EA 26.3 EUR
ROTHP034.1 ROTHP034.2 ROTHP034.5
Sodium thiosulphate pentahydrate
Sodium thiosulphate pentahydrate
Formula: Na₂S₂O₃.5H₂O
MW: 248.19 g/mol
Melting Pt: 48
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149186
CAS Number: 10102-17-7
EINECS: 231-867-5
Merck Index: 12,08844