Lithium sulphate monohydrate, Sigma-Aldrich®
Supplier: Merck
Warning
62612-100G 62612-500G
SIAL62612-100GEA 92 EUR
SIAL62612-100G SIAL62612-500G
Lithium sulphate monohydrate, Sigma-Aldrich®
Lithium sulphate monohydrate
Formula: Li₂SO₄·H₂O
MW: 127.96 g/mol
Melting Pt: 120 °C
Density: 2.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149766
CAS Number: 10102-25-7
EINECS: 233-820-4