Sodium sulphate decahydrate, Sigma-Aldrich®
Supplier: Merck
71969-1KG 71969-500G
SIAL71969-1KGEA 133 EUR
SIAL71969-1KG SIAL71969-500G
Sodium sulphate decahydrate, Sigma-Aldrich®
Sodium sulphate decahydrate
Formula: Na₂SO₄·10H₂O
MW: 322.2 g/mol
Boiling Pt: 1429
Melting Pt: 32.4
Density: 1.464 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149189
CAS Number: 7727-73-3
EINECS: 231-820-9
Merck Index: 13,08755