1-Iodooctane ≥98% stabilised
Supplier: Thermo Fisher Scientific
|
Warning
|
Stabilization:
Stabilised with copper
Stabilized with copper
Purity: 98+%
CAS No.: 629-27-6
Molecular Formula: C8H17I
Formula Weight: 240.13
Flash Point: 95°(203°F)
Refractive Index: 1.4878
Melting Point: -46°
Boiling Point: 225-226°
Beilstein Reference: 1697479
MDL No.: MFCD00001105
Formula:
C₈H₁₇I Boiling Pt: 225…226 °C Melting Pt: –46 °C Density: 1.325 Flash Pt: 95 °C (203 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001105 CAS Number: 629-27-6 EINECS: 211-084-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...