1,1,2,2-Tetrabromoethane, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ100
86760-1L
86760-250ML
SIAL86760-1LEA
351
EUR
SIAL86760-1L
SIAL86760-250ML
1,1,2,2-Tetrabromoethane, Sigma-Aldrich®
1,1,2,2-Tetrabromoethane
Formula:
Br₂HCCHBr₂ MW: 345.65 g/mol Boiling Pt: 244 °C (1013 hPa) Melting Pt: –1 °C Density: 2.9656 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000133 CAS Number: 79-27-6 EINECS: 201-191-5 UN: 2504 ADR: 6.1,III Merck Index: 13,09261 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...