1,10-Dibromodecane 97%
Supplier: Thermo Fisher Scientific
112780250 112781000 112785000
ACRO112780250EA 49.7 EUR
ACRO112780250 ACRO112781000 ACRO112785000
1,10-Dibromodecane 97%
1,10-Dibromodecane
Formula: BrH₂C(CH₂)₈CH₂Br
Boiling Pt: 160 °C (15 mmHg)
Melting Pt: 25…30 °C
Density: 1.34 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00000222
CAS Number: 4101-68-2
EINECS: 223-871-0
Order Now
Specification Test Results
Appearance White to light brown Fused solid or liquid
Infrared spectrum Conforms
Melting point 25 to 30 °C
GC ≥96.0 %