1,8-Dibromooctane
Supplier: Thermo Scientific
Synonyms: Octamethylendibromide
SB00705.1 SB00705.10 SB00705.50
MAYBSB00705.1EA 0 EUR
MAYBSB00705.1 MAYBSB00705.10 MAYBSB00705.50
1,8-Dibromooctane
1,8-Dibromooctane
Formula: BrH₂C(CH₂)₆CH₂Br
MW: 272.02 g/mol
Boiling Pt: 270
Melting Pt: 15
Density: 1.46 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000277
CAS Number: 4549-32-0
EINECS: 224-912-5