1-Bromooctadecane 96%
Supplier: Thermo Fisher Scientific
190021000
190025000
190020025
ACRO190021000EA
31.1
EUR
ACRO190021000
ACRO190025000
ACRO190020025
1-Bromooctadecane 96%
1-Bromooctadecane
Formula:
BrH₂C(CH₂)₁₆CH₃ |
MDL Number:
MFCD00000231 CAS Number: 112-89-0 |
Specification Test Results
Appearance | White to yellow Fused solid or liquid |
Infrared spectrum | Conforms |
Melting point | 25 to 31 °C |
GC | ≥95 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...