1-Bromo-3-chloropropane 99%
Supplier: Thermo Scientific
Danger
106860010 106862500 106860025 106860501
ACRO106860010EA 110 EUR
ACRO106860010 ACRO106862500 ACRO106860025 ACRO106860501
1-Bromo-3-chloropropane 99%
1-Bromo-3-chloropropane
Formula: BrH₂CCH₂CH₂Cl
MW: 157.44 g/mol
Boiling Pt: 142…145 °C (760 mmHg)
Melting Pt: –59 °C
Density: 1.59 g/cm³
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00000998
CAS Number: 109-70-6
EINECS: 203-697-1
UN: 2688
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4850 to 1.4880 (20°C, 589 nm)
GC ≥98.5 %
Color scale ≤50 APHA
Acidity (HCl) ≤0.002 %