Bromochloromethane, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
135267-100G
135267-2KG
SIAL135267-100GEA
19.2
EUR
SIAL135267-100G
SIAL135267-2KG
Bromochloromethane, Sigma-Aldrich®
Bromochloromethane
Formula:
BrClCH₂ Boiling Pt: 68…69 °C (1013 hPa) Melting Pt: –83 °C Density: 1.93 g/cm³ (20 °C) Storage Temperature: Ambient |
CAS Number:
74-97-5 EINECS: 200-826-3 UN: 1887 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...