Acetaldehyde ≥99.0%, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Ethanal
M-Clarity™ quality level = MQ200
00071-1L
00071-500ML
00071-5ML
SIAL00071-1LEA
163
EUR
SIAL00071-1L
SIAL00071-500ML
SIAL00071-5ML
Acetaldehyde ≥99.0%, Sigma-Aldrich®
Acetaldehyde
Formula:
CH₃CHO Boiling Pt: 20.1 °C (1013 hPa) Melting Pt: –123 °C Density: 0.78 g/cm³ (20 °C) Flash Pt: –27 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006991 CAS Number: 75-07-0 EINECS: 200-836-8 UN: 1089 ADR: 3,I Merck Index: 12,00037 |