Dispensers for KIMTECH* WETTASK* wipes
Supplier: Kimberly-Clark
Kimtech™ Wettask
115-2076EA 290 EUR
115-2076
Dispensers for KIMTECH* WETTASK* wipes
Wipe Dispensers
Dispenser buckets for KIMTECH* WETTASK* roll wipers. Bucket can easily be opened and refilled with wipes and sanitizers/solutions. This enclosed system reduces cross contamination, Volatile Organic Compounds and splashes.
  • Easy refilling with the appropriate solution
  • Pre-moistened cloths can easily be pulled through the aperture in the lid
  • Mobile usage: Portable