Propionaldehyde 99+%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Propanal
220511000 220510025 220515000
ACRO220511000EA 26 EUR
ACRO220511000 ACRO220510025 ACRO220515000
Propionaldehyde 99+%
Propionaldehyde
Formula: C₃H₆O
MW: 58.08 g/mol
Boiling Pt: 48 °C (760 mmHg)
Melting Pt: –81 °C
Density: 0.798 g/cm³
Flash Pt: –40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007020
CAS Number: 123-38-6
EINECS: 204-623-0
UN: 1275
ADR: 3 II
Merck Index: 13,07915
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3610 to 1.3660 (20°C, 589 nm)
GC ≥99.0 %
Color scale ≤20 APHA