Acetonitrile 99+%, extra pure
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
149520010 149520025 149520250 149520050
ACRO149520010EA 114 EUR
ACRO149520010 ACRO149520025 ACRO149520250 ACRO149520050
Acetonitrile 99+%, extra pure
Acetonitrile
Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81…82 °C (760 mmHg)
Melting Pt: –46 °C
Density: 0.781 g/cm³
Flash Pt: 2 °C
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3 II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear liquid
Water ≤0.3 % (K.F.)
GC ≥99.0 %
Color scale ≤20 APHA
Residue after evaporation ≤0.005 %