Acetonitrile ≥99%
Supplier: Thermo Fisher Scientific
|
Danger
|
Purity: 99%
CAS No.: 75-05-8
Molecular Formula: C2H3N
Formula Weight: 41.05
Flash Point: 5°(41°F)
Refractive Index: 1.3440
Odour: Pungent
Melting Point: -48°
Boiling Point: 80-82°
Beilstein Reference: 741857
Merck Reference: 14,70
UN No.: 1648
MDL No.: MFCD00001878
Formula:
H₃CCN Boiling Pt: 80…82 °C Melting Pt: –48 °C Density: 0.786 Flash Pt: 5 °C (41 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001878 CAS Number: 75-05-8 EINECS: 200-835-2 UN: 1648 ADR: 3,II REACH: 01-2119471307-38 Merck Index: 14,70 |