(1R)-(-)-Camphor-10-sulphonic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: L(-)-Camphor-10-sulphonic acid, (1R)-(-)-2-Oxobornane-10-sulphonic acid, ((1R,4S)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulphonic acid, (-)-CSA
MQ100
282146-100G 282146-25G
SIAL282146-100GEA 98.6 EUR
SIAL282146-100G SIAL282146-25G
(1R)-(-)-Camphor-10-sulphonic acid, Sigma-Aldrich®
(1R)-(-)-Camphor-10-sulphonic acid
Formula: C₁₀H₁₆O₄S
MW: 232.3 g/mol
Boiling Pt: 375
Melting Pt: 193…196 °C
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00150753
CAS Number: 35963-20-3
EINECS: 252-817-9