(1S)-(+)-Camphor-10-sulphonyl chloride, Sigma-Aldrich®
Supplier: Merck
Synonyms: (1S)-(+)-2-Oxobornane-10-sulphonyl chloride, ((1S,4R)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulphonyl chloride, D(+)-Camphor-10-sulphonyl chloride
MQ200
219576-100G 219576-25G 219576-5G
SIAL219576-100GEA 432 EUR
SIAL219576-100G SIAL219576-25G SIAL219576-5G
(1S)-(+)-Camphor-10-sulphonyl chloride, Sigma-Aldrich®
(1S)-(+)-Camphor-10-sulphonyl chloride
Formula: C₁₀H₁₅ClO₃S
MW: 250.75
Melting Pt: 70
MDL Number: MFCD00064156
CAS Number: 21286-54-4
EINECS: 244-314-8
UN: 3261
ADR: 8 II