D-Glucono-1,5-lactone, Sigma-Aldrich®
Supplier: Merck
Synonyms: GDL, D-Glucono-δ-lactone, D-Gluconic acid-1,5-lactone, delta-Gluconolactone, D-Gluconic acid lactone, (3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-one, D-(+)-Glucono-δ-lactone, D-(+)-Gluconic acid δ-lactone, D-Gluconic acid-δ-lacton, D-(+)-Glucono-1,5-lactone, Gluconolactone
MQ200
G2164-100G G2164-1KG
SIALG2164-100GEA 40.9 EUR
SIALG2164-100G SIALG2164-1KG
D-Glucono-1,5-lactone, Sigma-Aldrich®
D-Glucono-1,5-lactone
Formula: C₆H₁₀O₆
MW: 178.14 g/mol
Boiling Pt: 385
Melting Pt: 153
Density: 1.61 g/cm³ (–5 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006647
CAS Number: 90-80-2
EINECS: 202-016-5
Merck Index: 13,04470