3-Phenylpropylamine, Sigma-Aldrich®
Supplier: Merck
Synonyms:
3-Phenyl-1-propylamine
M-Clarity™ quality level = MQ200
P32406-10G
P32406-25G
SIALP32406-10GEA
46.4
EUR
SIALP32406-10G
SIALP32406-25G
3-Phenylpropylamine, Sigma-Aldrich®
3-Phenylpropylamine
Formula:
C₉H₁₃N MW: 135.21 g/mol Boiling Pt: 221 °C (1013 hPa) Density: 0.951 g/cm³ (15 °C) |
MDL Number:
MFCD00008224 CAS Number: 2038-57-5 EINECS: 218-012-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...