3-Phenyl-2-propyn-1-ol, Sigma-Aldrich®

Supplier: Merck

Synonyms: 3-Phenylprop-2-yn-1-ol

M-Clarity™ quality level = MQ100

589306-25G 589306-5G
SIAL589306-25GEA 107 EUR
SIAL589306-25G SIAL589306-5G
3-Phenyl-2-propyn-1-ol, Sigma-Aldrich®
3-Phenyl-2-propyn-1-ol
Formula: C₉H₈O
MW: 132.16 g/mol
MDL Number: MFCD00040914
CAS Number: 1504-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR