(3-Pyridyl)acetic acid ethyl ester, Sigma-Aldrich®

Supplier: Merck

Synonyms: Ethyl 3-pyridineacetate, Ethyl 2-(pyridin-3-yl)acetate, Ethyl (3-pyridyl)acetate, Ethyl 3-pyridylacetate

M-Clarity™ quality level = MQ100

E47255-25G E47255-5G
SIALE47255-25GEA 296 EUR
SIALE47255-25G SIALE47255-5G
(3-Pyridyl)acetic acid ethyl ester, Sigma-Aldrich®
(3-Pyridyl)acetic acid ethyl ester
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 80 °C (1013 hPa)
Density: 1.08 g/cm³ (20 °C)
MDL Number: MFCD00006411
CAS Number: 39931-77-6
EINECS: 254-707-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR