(4-Carboxybutyl)triphenylphosphonium bromide, Sigma-Aldrich®

Supplier: Merck

Synonyms: (4-Carboxybut-1-yl)(triphenyl)phosphonium bromide

M-Clarity™ quality level = MQ200

157945-250G 157945-50G
SIAL157945-250GEA 329 EUR
SIAL157945-250G SIAL157945-50G
(4-Carboxybutyl)triphenylphosphonium bromide, Sigma-Aldrich®
(4-Carboxybutyl)triphenylphosphonium bromide
Formula: C₂₃H₂₄BrO₂P
MW: 443.32 g/mol
Melting Pt: 205…208 °C
MDL Number: MFCD00011906
CAS Number: 17814-85-6
EINECS: 241-782-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR