N-Methylaniline, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M29304-1KG
M29304-25G
M29304-500G
SIALM29304-1KGEA
83.2
EUR
SIALM29304-1KG
SIALM29304-25G
SIALM29304-500G
N-Methylaniline, Sigma-Aldrich®
N-Methylaniline
Formula:
C₇H₉N MW: 107.16 g/mol Boiling Pt: 196.3 °C (1013 hPa) Melting Pt: –57 °C Density: 0.989 g/cm³ (20 °C) Flash Pt: 84 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008283 CAS Number: 100-61-8 EINECS: 202-870-9 UN: 2294 ADR: 6.1,III Merck Index: 12,06098 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...