1,4-Dithioerythritol, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
D8161-10G D8161-1G D8161-50G D8161-5G
SIALD8161-10GEA 428 EUR
SIALD8161-10G SIALD8161-1G SIALD8161-50G SIALD8161-5G
1,4-Dithioerythritol, Sigma-Aldrich®
1,4-Dithioerythritol
Formula: C₄H₁₀O₂S₂
Boiling Pt: 286 °C (1013 hPa)
Melting Pt: 82…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00063750
CAS Number: 6892-68-8
EINECS: 229-998-8