(S)-(+)-Carvone, Sigma-Aldrich®

Supplier: Merck

Synonyms: D-p-Mentha-1(6),8-dien-2-one, D(+)-Carvone, (S)-2-Methyl-5-(prop-1-en-2-yl)cyclohex-2-enone, (S)-(+)-2-Methyl-5-(1-methylvinyl)cyclohex-2-en-1-one

M-Clarity™ quality level = MQ100

435759-25ML 435759-5ML
SIAL435759-25MLEA 121 EUR
SIAL435759-25ML SIAL435759-5ML
(S)-(+)-Carvone, Sigma-Aldrich®
(S)-(+)-Carvone
Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: ? 15 °C
Density: 0.9584 g/cm³ (20 °C)
MDL Number: MFCD00062997
CAS Number: 2244-16-8
EINECS: 218-827-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR