Triphosgene, Sigma-Aldrich®
Supplier: Merck
Synonyms: Bis(trichloromethyl) carbonate
15217-10G 15217-50G
SIAL15217-10GEA 101 EUR
SIAL15217-10G SIAL15217-50G
Triphosgene, Sigma-Aldrich®
Triphosgene
Formula: C₃Cl₆O₃
MW: 296.75 g/mol
Boiling Pt: 203
Melting Pt: 77…81 °C
Density: 1.629 g/cm³ (80 °C)
MDL Number: MFCD00062848
CAS Number: 32315-10-9
EINECS: 250-986-3
UN: 2928
ADR: 6.1 II