α,α,α-Trifluoroacetophenone
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2,2,2-Trifluoroacetophenone, Phenacyl trifluoride, Trifluoroacetylbenzene, 1-Phenyl-2,2,2-trifluoro-1-ethanone
SB01827.1
SB01827.10
SB01827.50
MAYBSB01827.1EA
0
EUR
MAYBSB01827.1
MAYBSB01827.10
MAYBSB01827.50
α,α,α-Trifluoroacetophenone
α,α,α-Trifluoroacetophenone
Formula:
C₈H₅F₃O MW: 174.12 g/mol Boiling Pt: 165 °C (1013 hPa) Melting Pt: –40 °C Density: 1.2799 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000420 CAS Number: 434-45-7 EINECS: 207-103-1 UN: 1224 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...