Trimethyl orthobenzoate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Trimethyl orthobeznoate, (Trimethoxymethyl)benzene
M-Clarity™ quality level = MQ100
164534-10G 164534-50G
SIAL164534-10GEA 86.1 EUR
SIAL164534-10G SIAL164534-50G
Trimethyl orthobenzoate, Sigma-Aldrich®
Trimethyl orthobenzoate
Formula: C₁₀H₁₄O₃
MW: 182.22 g/mol
MDL Number: MFCD00008474
CAS Number: 707-07-3
EINECS: 211-897-5