Dopamine hydrochloride, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1-(2-Aminoethyl)-3,4-dihydroxybenzene HCl, 3-Hydroxytyraminium chloride, 3-Hydroxytyramine hydrochloride, 3,4-Dihydroxyphenethylammonium chloride
MQ200
H8502-5G H8502-100G H8502-10G
SIALH8502-5GEA 40.9 EUR
SIALH8502-5G SIALH8502-100G SIALH8502-10G
Dopamine hydrochloride, Sigma-Aldrich®
Dopamine hydrochloride
Formula: C₈H₁₂ClNO₂
MW: 189.64
Melting Pt: 246…249 °C
Density: 1.356 g/cm³ (20 °C)
MDL Number: MFCD00012898
CAS Number: 62-31-7
EINECS: 200-527-8