Tris(perfluorobutyl)amine, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Perfluorotri-n-butylamine, Heptacosafluorotri-n-butylamine, Perfluorotributylamine, FC-43, Heptacosafluorotributylamine, PFTBA, Tris(nonafluorobutyl)amine
M-Clarity™ quality level = MQ200
H5262-25ML
H5262-5ML
SIALH5262-25MLEA
686
EUR
SIALH5262-25ML
SIALH5262-5ML
Tris(perfluorobutyl)amine, Sigma-Aldrich®
Tris(perfluorobutyl)amine
Formula:
C₁₂F₂₇N Boiling Pt: 155 °C (1013 hPa) Melting Pt: –55 °C Density: 1.883 g/cm³ (20 °C) |
MDL Number:
MFCD00000436 CAS Number: 311-89-7 EINECS: 206-223-1 |