4-(Dimethylamino)benzophenone, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
149349-10G
149349-25G
SIAL149349-10GEA
169
EUR
SIAL149349-10G
SIAL149349-25G
4-(Dimethylamino)benzophenone, Sigma-Aldrich®
4-(Dimethylamino)benzophenone
Formula:
C₁₅H₁₅NO MW: 225.29 g/mol Boiling Pt: 245…250 °C (15 mmHg) Melting Pt: 97…98 °C |
MDL Number:
MFCD00008311 CAS Number: 530-44-9 EINECS: 208-478-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...